Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260558.2 | NP_001247487.1 | 1366 | 70 | ||
XM_015110330.2 | XP_014965816.1 | 1657 | 70 | |||
GenBank mRNA | JU327771.1 | - | 1358 | 70 | ||
JV046272.1 | - | 1371 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |