Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS26B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257745.1 | NP_001244674.1 | 992 | 97 | ||
GenBank mRNA | JU320309.1 | - | 992 | 97 | ||
JU473342.1 | - | 855 | 97 | |||
JV044752.1 | - | 722 | 97 | |||
JV667667.1 | - | 1005 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |