Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015110934.2 | XP_014966420.2 | 494 | 54 | ||
XM_015110935.2 | XP_014966421.2 | 929 | 54 | |||
GenBank mRNA | JU327317.1 | - | 641 | 54 | ||
JV060016.1 | - | 764 | 54 | |||
JV650362.1 | - | 765 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |