Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194766.2 | NP_001181695.1 | 292 | 83 | ||
XM_015152920.1 | XP_015008406.1 | 292 | 83 | |||
GenBank mRNA | JU320504.1 | - | 226 | 83 | ||
JU474110.1 | - | 269 | 83 | |||
JV044838.1 | - | 297 | 83 | |||
JV601370.1 | - | 279 | 83 |