Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261338.1 | NP_001248267.1 | 518 | 80 | ||
XM_015141895.2 | XP_014997381.1 | 924 | 80 | |||
XM_015141896.2 | XP_014997382.1 | 562 | 80 | |||
XM_015141898.2 | XP_014997384.1 | 493 | 80 | |||
XM_015141899.2 | XP_014997385.1 | 497 | 80 | |||
XM_015141900.2 | XP_014997386.1 | 526 | 80 | |||
GenBank mRNA | JU336916.1 | - | 507 | 80 | ||
JU336917.1 | - | 507 | 80 | |||
JU477651.1 | - | 520 | 80 | |||
JV048584.1 | - | 518 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |