Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260730.1 | NP_001247659.1 | 456 | 117 | ||
XM_015142229.2 | XP_014997715.2 | 600 | 117 | |||
XM_015142232.2 | XP_014997718.2 | 603 | 117 | |||
XM_028850495.1 | XP_028706328.1 | 978 | 117 | |||
GenBank mRNA | JU322502.1 | - | 543 | 117 | ||
JU476475.1 | - | 456 | 117 | |||
JV047713.1 | - | 521 | 117 | |||
JV656460.1 | - | 546 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |