Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGFRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015136684.2 | XP_014992170.1 | 986 | 160 | ||
GenBank mRNA | JU323158.1 | - | 463 | 160 | ||
JV723371.1 | - | 515 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |