Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260729.1 | NP_001247658.1 | 289 | 87 | ||
XM_015142767.2 | XP_014998253.1 | 141 | 87 | |||
GenBank mRNA | JU332405.1 | - | 462 | 87 | ||
JU473866.1 | - | 281 | 87 | |||
JV045838.1 | - | 289 | 87 | |||
JV491365.1 | - | 341 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |