Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266979.1 | NP_001253908.1 | 402 | 63 | ||
GenBank mRNA | JU322341.1 | - | 402 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |