Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261300.1 | NP_001248229.1 | 448 | 106 | ||
XM_028845413.1 | XP_028701246.1 | 641 | 106 | |||
GenBank mRNA | JU332453.1 | - | 541 | 106 | ||
JU473920.1 | - | 542 | 106 | |||
JV045061.1 | - | 448 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |