Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260546.2 | NP_001247475.1 | 576 | 119 | ||
GenBank mRNA | JU328023.1 | - | 339 | 119 | ||
JV046964.1 | - | 579 | 119 | |||
JV635446.1 | - | 814 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |