Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAZL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028843651.1 | XP_028699484.1 | 1027 | 128 | ||
GenBank mRNA | JV044123.1 | - | 973 | 128 | ||
JV044124.1 | - | 1104 | 128 | |||
JV637401.1 | - | 1052 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |