Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AUH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257675.1 | NP_001244604.1 | 456 | 119 | ||
XM_015118074.2 | XP_014973560.2 | 534 | 119 | |||
GenBank mRNA | JU335142.1 | - | 456 | 119 | ||
JV047465.1 | - | 433 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |