Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BEND5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266827.2 | NP_001253756.1 | 909 | 85 | ||
XM_015139078.2 | XP_014994564.1 | 1151 | 85 | |||
XM_015139089.2 | XP_014994575.1 | 1117 | 85 | |||
XM_028844223.1 | XP_028700056.1 | 1495 | 85 | |||
XM_028844227.1 | XP_028700060.1 | 1459 | 85 | |||
XM_028844233.1 | XP_028700066.1 | 1497 | 85 | |||
XM_028844260.1 | XP_028700093.1 | 1334 | 85 | |||
XM_028844276.1 | XP_028700109.1 | 1225 | 85 | |||
GenBank mRNA | JU328464.1 | - | 758 | 85 | ||
JV635600.1 | - | 911 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |