Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUCKS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266481.1 | NP_001253410.1 | 145 | 90 | ||
XM_015118335.2 | XP_014973821.1 | 321 | 90 | |||
GenBank mRNA | JU319617.1 | - | 153 | 90 | ||
JU319618.1 | - | 153 | 90 | |||
JU471457.1 | - | 258 | 90 | |||
JV625906.1 | - | 305 | 90 | |||
JV635557.1 | - | 145 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |