Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001113452.4 | XP_001113452.1 | 1573 | 64 | ||
XM_015149141.2 | XP_015004627.1 | 1101 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |