Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB27B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001083132.4 | XP_001083132.1 | 501 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |