Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082091.4 | XP_001082091.1 | 903 | 87 | ||
XM_001082213.4 | XP_001082213.1 | 915 | 87 | |||
XM_028828543.1 | XP_028684376.1 | 1106 | 87 | |||
XM_028828544.1 | XP_028684377.1 | 1106 | 87 | |||
GenBank mRNA | JV708728.1 | - | 223 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |