Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL6R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001114404.4 | XP_001114404.2 | 1649 | 111 | ||
GenBank mRNA | JU485887.1 | - | 902 | 111 | ||
JV060090.1 | - | 328 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |