Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXPH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266633.2 | NP_001253562.1 | 184 | 75 | ||
GenBank mRNA | JU321065.1 | - | 184 | 75 | ||
JV637775.1 | - | 208 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |