Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194054.1 | NP_001180983.1 | 495 | 64 | ||
XM_015110985.2 | XP_014966471.1 | 959 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |