Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST6GALNAC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001102189.4 | XP_001102189.1 | 647 | 77 | ||
XM_015142889.2 | XP_014998375.1 | 647 | 77 | |||
GenBank mRNA | JU326427.1 | - | 80 | 77 | ||
JV600322.1 | - | 397 | 77 | |||
JV724413.1 | - | 445 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |