Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001085740.4 | XP_001085740.4 | 457 | 99 | ||
GenBank mRNA | JU335512.1 | - | 368 | 99 | ||
JU476452.1 | - | 367 | 99 | |||
JV066077.1 | - | 350 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |