Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001092669.4 | XP_001092669.1 | 579 | 71 | ||
GenBank mRNA | JU495427.1 | - | 187 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta Inventoried |