Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194768.3 | NP_001181697.1 | 302 | 108 | ||
XM_001098260.4 | XP_001098260.1 | 587 | 108 | |||
XM_015110255.2 | XP_014965741.1 | 564 | 108 | |||
XM_015110256.2 | XP_014965742.1 | 172 | 108 | |||
XM_015110258.2 | XP_014965744.1 | 414 | 108 | |||
XM_015110259.2 | XP_014965745.1 | 1365 | 108 | |||
XM_028830552.1 | XP_028686385.1 | 568 | 108 | |||
GenBank mRNA | JU322139.1 | - | 259 | 108 | ||
JU473186.1 | - | 258 | 108 | |||
JV605976.1 | - | 262 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |