Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPLL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112137.2 | XP_014967623.1 | 1107 | 67 | ||
XM_015112138.2 | XP_014967624.1 | 1203 | 67 | |||
XM_015112139.2 | XP_014967625.1 | 957 | 67 | |||
XM_015112144.2 | XP_014967630.1 | 1115 | 67 | |||
GenBank mRNA | JU320592.1 | - | 864 | 67 | ||
JU471277.1 | - | 1209 | 67 | |||
JU471278.1 | - | 947 | 67 | |||
JV048711.1 | - | 864 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |