Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNTB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001100304.4 | XP_001100304.2 | 2138 | 61 | ||
GenBank mRNA | JU492038.1 | - | 1109 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |