Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261792.1 | NP_001248721.1 | 961 | 55 | ||
GenBank mRNA | JU337139.1 | - | 961 | 55 | ||
JV526004.1 | - | 346 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |