Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001111920.3 | XP_001111920.1 | 2266 | 71 | ||
XM_001111958.3 | XP_001111958.1 | 1879 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |