Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JADE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266133.1 | NP_001253062.1 | 217 | 62 | ||
XM_015138933.2 | XP_014994419.1 | 98 | 62 | |||
XM_015138934.2 | XP_014994420.1 | 469 | 62 | |||
XM_015138935.2 | XP_014994421.1 | 206 | 62 | |||
XM_015138936.2 | XP_014994422.1 | 288 | 62 | |||
XM_028847943.1 | XP_028703776.1 | 180 | 62 | |||
XM_028847944.1 | XP_028703777.1 | 213 | 62 | |||
GenBank mRNA | JU336457.1 | - | 180 | 62 | ||
JU477273.1 | - | 183 | 62 | |||
JV048293.1 | - | 217 | 62 | |||
JV661899.1 | - | 311 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |