Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IKZF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261672.1 | NP_001248601.1 | 154 | 131 | ||
GenBank mRNA | JU335916.1 | - | 232 | 131 | ||
JU472153.1 | - | 250 | 131 | |||
JV044623.1 | - | 154 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |