Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193860.2 | NP_001180789.1 | 263 | 126 | ||
XM_015145400.2 | XP_015000886.1 | 532 | 126 | |||
XM_028852147.1 | XP_028707980.1 | 532 | 126 | |||
GenBank mRNA | JU319780.1 | - | 398 | 126 | ||
JU470671.1 | - | 306 | 126 | |||
JV047810.1 | - | 398 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |