Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3GL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266277.1 | NP_001253206.1 | 376 | 112 | ||
XM_015150043.2 | XP_015005529.1 | 531 | 112 | |||
XM_015150050.2 | XP_015005536.1 | 790 | 112 | |||
XM_028831801.1 | XP_028687634.1 | 933 | 112 | |||
XM_028831832.1 | XP_028687665.1 | 1054 | 112 | |||
XM_028831924.1 | XP_028687757.1 | 735 | 112 | |||
GenBank mRNA | JU321708.1 | - | 429 | 112 | ||
JV045313.1 | - | 376 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |