Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TADA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266137.1 | NP_001253066.1 | 736 | 83 | ||
GenBank mRNA | JU332518.1 | - | 728 | 83 | ||
JU473995.1 | - | 920 | 83 | |||
JV045919.1 | - | 736 | 83 | |||
JV452113.1 | - | 606 | 83 | |||
JV640245.1 | - | 2229 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |