Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194106.1 | NP_001181035.1 | 499 | 101 | ||
XM_028841782.1 | XP_028697615.1 | 510 | 101 | |||
GenBank mRNA | JU322812.1 | - | 399 | 101 | ||
JU475866.1 | - | 399 | 101 | |||
JV508990.1 | - | 402 | 101 | |||
JV653154.1 | - | 421 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |