Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFTPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257727.1 | NP_001244656.1 | 2165 | 67 | ||
XM_015112495.2 | XP_014967981.1 | 2230 | 67 | |||
XM_015112496.2 | XP_014967982.1 | 1985 | 67 | |||
GenBank mRNA | JU332285.1 | - | 2165 | 67 | ||
JU332286.1 | - | 2165 | 67 | |||
JU472339.1 | - | 2220 | 67 | |||
JU472340.1 | - | 2220 | 67 | |||
JV044200.1 | - | 2146 | 67 | |||
JV566312.1 | - | 2229 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |