Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193777.2 | NP_001180706.1 | 423 | 90 | ||
GenBank mRNA | JU320843.1 | - | 391 | 90 | ||
JU473650.1 | - | 350 | 90 | |||
JV044150.1 | - | 343 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |