Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPDU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266148.1 | NP_001253077.1 | 121 | 80 | ||
XM_015118493.2 | XP_014973979.1 | 148 | 80 | |||
XM_015118494.2 | XP_014973980.1 | 173 | 80 | |||
XM_015118495.2 | XP_014973981.1 | 325 | 80 | |||
GenBank mRNA | JU337208.1 | - | 121 | 80 | ||
JV637711.1 | - | 251 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |