Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266315.1 | NP_001253244.1 | 1418 | 77 | ||
XM_015140869.2 | XP_014996355.1 | 1536 | 77 | |||
XM_015140871.2 | XP_014996357.1 | 1611 | 77 | |||
XM_015140872.2 | XP_014996358.1 | 2106 | 77 | |||
XM_028849266.1 | XP_028705099.1 | 1510 | 77 | |||
GenBank mRNA | JU334508.1 | - | 1514 | 77 | ||
JU334509.1 | - | 1514 | 77 | |||
JU334510.1 | - | 1514 | 77 | |||
JU334511.1 | - | 1514 | 77 | |||
JU334512.1 | - | 1514 | 77 | |||
JU334513.1 | - | 1514 | 77 | |||
JU334514.1 | - | 1514 | 77 | |||
JU472412.1 | - | 1418 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |