Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266345.1 | NP_001253274.1 | 1022 | 87 | ||
GenBank mRNA | JU328471.1 | - | 707 | 87 | ||
JU477267.1 | - | 1017 | 87 | |||
JU477268.1 | - | 1017 | 87 | |||
JU477269.1 | - | 1017 | 87 | |||
JU477270.1 | - | 1017 | 87 | |||
JU477271.1 | - | 1017 | 87 | |||
JV048292.1 | - | 1022 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |