Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115137.2 | NP_001108609.2 | 398 | 77 | ||
GenBank mRNA | CO774353.1 | - | 281 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta Inventoried |