Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC4F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112720.2 | XP_014968206.2 | 268 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |