Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001084475.4 | XP_001084475.3 | 1075 | 129 | ||
XM_028850693.1 | XP_028706526.1 | 1076 | 129 | |||
XM_028850694.1 | XP_028706527.1 | 1080 | 129 | |||
GenBank mRNA | JU323442.1 | - | 636 | 129 | ||
JU473039.1 | - | 921 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |