Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032859.1 | NP_001028031.1 | 1215 | 70 | ||
XM_015120210.2 | XP_014975696.1 | 1611 | 70 | |||
GenBank mRNA | AF021135.1 | - | 1215 | 70 | ||
JU334421.1 | - | 1593 | 70 | |||
JU334422.1 | - | 1593 | 70 | |||
JU334423.1 | - | 1593 | 70 | |||
JU334424.1 | - | 1593 | 70 | |||
JU475515.1 | - | 1431 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |