Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032945.1 | NP_001028117.1 | 1061 | 105 | ||
XM_015113701.2 | XP_014969187.1 | 1795 | 105 | |||
XM_015113702.2 | XP_014969188.1 | 1226 | 105 | |||
XM_015113703.2 | XP_014969189.1 | 1319 | 105 | |||
GenBank mRNA | AF026262.1 | - | 1061 | 105 | ||
JU332769.1 | - | 1255 | 105 | |||
JV604769.1 | - | 1409 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Macaca mulatta |