Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193827.3 | NP_001180756.2 | 523 | 84 | ||
XM_028840350.1 | XP_028696183.1 | 624 | 84 | |||
GenBank mRNA | AY680580.1 | - | 122 | 84 | ||
JV605230.1 | - | 576 | 84 |