Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032963.1 | NP_001028135.1 | 438 | 96 | ||
GenBank mRNA | M83999.1 | - | 438 | 96 | ||
S40060.1 | - | 438 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |