Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKG2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032889.1 | NP_001028061.1 | 295 | 129 | ||
XM_015150990.2 | XP_015006476.2 | 470 | 129 | |||
GenBank mRNA | AF190943.1 | - | 295 | 129 | ||
AJ554302.1 | - | 282 | 129 | |||
JU474751.1 | - | 423 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |