Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF190983.1 | - | 220 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |