Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYROBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032867.1 | NP_001028039.1 | 270 | 92 | ||
XM_015123562.2 | XP_014979048.1 | 467 | 92 | |||
GenBank mRNA | AF321611.1 | - | 270 | 92 | ||
JU322380.1 | - | 298 | 92 | |||
JU473718.1 | - | 291 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |